accessibility menu, dialog, popup

UserName

BrN

CAS RN 5332-25-2

IUPAC Name: 6-bromoquinoline
Synonyms: 6-bromoquinoline
Molecular Weight (g/mol): 208.06
Molecular Formula: C9H6BrN
InChi Key: IFIHYLCUKYCKRH-UHFFFAOYSA-N
SMILES: BrC1=CC=C2N=CC=CC2=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)