missing translation for 'onlineSavingsMsg'
Learn More

6-Bromoquinoline, 97%

Catalog No. p-7036753
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5511103 1 g
AAH5511106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5511103 Supplier Thermo Scientific Chemicals Supplier No. H5511103

CAS: 5332-25-2 | C9H6BrN | 208.058 g/mol

6-Bromoquinoline is used as fine chemical and pharmaceutical intermediate, used as the coupling reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromoquinoline is used as fine chemical and pharmaceutical intermediate, used as the coupling reagent.

Solubility
Soluble in acetone, acetonitrile, dichloromethane, ethyl acetate and THF.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 5332-25-2
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.058
MDL Number MFCD00024023
InChI Key IFIHYLCUKYCKRH-UHFFFAOYSA-N
Synonym quinoline, 6-bromo, 6-bromo-quinoline, 6-bromo quinoline, 6-bromchinolin, 6-bromchinolin;, 6-br-quinoline, 6-bromooquinoline, acmc-20aits, pubchem2374, 6-bromoquinoline
PubChem CID 79243
IUPAC Name 6-bromoquinoline
SMILES C1=CC2=C(C=CC(=C2)Br)N=C1

Specifications

Melting Point 24°C
Density 1.538
Flash Point >110°C (230°F)
Refractive Index 1.663
Quantity 1 g
Solubility Information Soluble in acetone,acetonitrile,dichloromethane,ethyl acetate and THF.
Formula Weight 208.06
Percent Purity 97%
Chemical Name or Material 6-Bromoquinoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 226-238-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only