Learn More
6-Bromopyridine-2-carboxylic acid, 97%
CAS: 21190-87-4 | C6H4BrNO2 | 202.007 g/mol
Supplier: Thermo Scientific Chemicals H3055306
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 6-Bromopyridine-2-carboxylic acid | |
| ∽188°C (decomposition) | |
| MFCD00093197 | |
| 6-bromopicolinic acid, 6-bromo-2-pyridinecarboxylic acid, 6-bromo-2-pyridine carboxylic acid, 2-pyridinecarboxylic acid, 6-bromo, 2-bromo pyridine-6-carboxylic acid, 2-bromopyridine-6-carboxylic acid, 2-bromo-6-pyridinecarboxylic acid, 6-bromo-pyridine-2-carboxylic acid, 6-bromopicolinicacid, pubchem2422 | |
| C1=CC(=NC(=C1)Br)C(=O)O | |
| 202.007 | |
| 202.01 |
| 21190-87-4 | |
| C6H4BrNO2 | |
| 5 g | |
| XURXQNUIGWHWHU-UHFFFAOYSA-N | |
| 6-bromopyridine-2-carboxylic acid | |
| 593919 | |
| 97% |
Chemical Identifiers
| 21190-87-4 | |
| 202.007 | |
| XURXQNUIGWHWHU-UHFFFAOYSA-N | |
| 593919 | |
| C1=CC(=NC(=C1)Br)C(=O)O |
| C6H4BrNO2 | |
| MFCD00093197 | |
| 6-bromopicolinic acid, 6-bromo-2-pyridinecarboxylic acid, 6-bromo-2-pyridine carboxylic acid, 2-pyridinecarboxylic acid, 6-bromo, 2-bromo pyridine-6-carboxylic acid, 2-bromopyridine-6-carboxylic acid, 2-bromo-6-pyridinecarboxylic acid, 6-bromo-pyridine-2-carboxylic acid, 6-bromopicolinicacid, pubchem2422 | |
| 6-bromopyridine-2-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only