accessibility menu, dialog, popup

UserName

OOBrN

CAS RN 21190-87-4

IUPAC Name: 6-bromopyridine-2-carboxylate
Synonyms: 6-bromopyridine-2-carboxylate
Molecular Weight (g/mol): 201
Molecular Formula: C6H3BrNO2
InChi Key: XURXQNUIGWHWHU-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=CC(Br)=N1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)