missing translation for 'onlineSavingsMsg'
Learn More

6-Bromopyridine-2-carboxylic acid, 97%

Catalog No. AAH3055303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3055303 1 g
AAH3055306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3055303 Supplier Thermo Scientific Chemicals Supplier No. H3055303
Only null left

CAS: 21190-87-4 | C6H4BrNO2 | 202.007 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21190-87-4
Molecular Formula C6H4BrNO2
Molecular Weight (g/mol) 202.007
MDL Number MFCD00093197
InChI Key XURXQNUIGWHWHU-UHFFFAOYSA-N
Synonym 6-bromopicolinic acid, 6-bromo-2-pyridinecarboxylic acid, 6-bromo-2-pyridine carboxylic acid, 2-pyridinecarboxylic acid, 6-bromo, 2-bromo pyridine-6-carboxylic acid, 2-bromopyridine-6-carboxylic acid, 2-bromo-6-pyridinecarboxylic acid, 6-bromo-pyridine-2-carboxylic acid, 6-bromopicolinicacid, pubchem2422
PubChem CID 593919
IUPAC Name 6-bromopyridine-2-carboxylic acid
SMILES C1=CC(=NC(=C1)Br)C(=O)O

Specifications

Melting Point ∽188°C (decomposition)
Quantity 1 g
Formula Weight 202.01
Percent Purity 97%
Chemical Name or Material 6-Bromopyridine-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only