missing translation for 'onlineSavingsMsg'
Learn More

6-Bromopiperonal, 98%

Catalog No. p-7023695
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1751006 5 g
AAA1751014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1751006 Supplier Thermo Scientific Chemicals Supplier No. A1751006
Only null left

CAS: 15930-53-7 | C8H5BrO3 | 229.03 g/mol

6-Bromopiperonal is used as an intermediate in the preparation of GPR30-selective agonist G-1.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromopiperonal is used as an intermediate in the preparation of GPR30-selective agonist G-1.

Solubility
Soluble in DMSO, methanol

Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 15930-53-7
Molecular Formula C8H5BrO3
Molecular Weight (g/mol) 229.03
MDL Number MFCD00022952
InChI Key CSQUXTSIDQURDV-UHFFFAOYSA-N
Synonym 6-bromopiperonal, 6-bromo-benzo 1,3 dioxole-5-carbaldehyde, 2-bromo-4,5-methylenedioxybenzaldehyde, piperonal, 6-bromo, 6-bromobenzo d 1,3 dioxole-5-carbaldehyde, 6-bromo-1,3-benzodioxole-5-carboxaldehyde, 6-bromo-2h-1,3-benzodioxole-5-carbaldehyde, 1,3-benzodioxole-5-carboxaldehyde, 6-bromo, 6-bromobenzo 1,3 dioxole-5-carbaldehyde, benzaldehyde, 2-bromo-4,5-methylenedioxy
PubChem CID 95062
SMILES BrC1=CC2=C(OCO2)C=C1C=O

Specifications

Melting Point 127°C to 130°C
Quantity 5 g
Sensitivity Air Sensitive
Solubility Information Soluble in DMSO,methanol
IUPAC Name 6-bromo-1,3-benzodioxole-5-carbaldehyde
Formula Weight 229.03
Percent Purity 98%
Chemical Name or Material 6-Bromopiperonal
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only