accessibility menu, dialog, popup

UserName

CAS RN 15930-53-7

IUPAC Name: 6-bromo-2H-1,3-benzodioxole-5-carbaldehyde
Synonyms: 6-bromo-2H-1,3-benzodioxole-5-carbaldehyde
Molecular Weight (g/mol): 229.03
Molecular Formula: C8H5BrO3
InChi Key: CSQUXTSIDQURDV-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(OCO2)C=C1C=O
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (1)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)