missing translation for 'onlineSavingsMsg'
Learn More

6-Bromophthalazin-1(4H)-one, 98%

Catalog No. p-7056597
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6560903 1 g
AAH6560906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6560903 Supplier Thermo Scientific Chemicals Supplier No. H6560903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 75884-70-7 | C8H5BrN2O | 225.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75884-70-7
Molecular Formula C8H5BrN2O
Molecular Weight (g/mol) 225.05
MDL Number MFCD09264003
InChI Key QMONLZVJOOMKRW-UHFFFAOYSA-N
PubChem CID 11535918
IUPAC Name 6-bromo-1,2-dihydrophthalazin-1-one
SMILES BrC1=CC=C2C(=O)NN=CC2=C1

Specifications

Quantity 1 g
Beilstein 10426132
Synonym 6-bromophthalazin-1 2h-one, 6-bromophthalazin-1-ol, 6-bromo-1,2-dihydrophthalazin-1-one, 1 2h-phthalazinone,6-bromo, 1 2h-phthalazinone, 6-bromo, ksc642c5d, 6-bromophthalazine-1 2h-one, 6-bromanyl-2h-phthalazin-1-one
Formula Weight 225.04
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 6-Bromophthalazin-1(4H)-one
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only