accessibility menu, dialog, popup

UserName

CAS RN 75884-70-7

IUPAC Name: 6-bromo-1,2-dihydrophthalazin-1-one
Synonyms: 6-bromo-2H-phthalazin-1-one
Molecular Weight (g/mol): 225.05
Molecular Formula: C8H5BrN2O
InChi Key: QMONLZVJOOMKRW-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C(=O)NN=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results