missing translation for 'onlineSavingsMsg'
Learn More

6-Bromooxindole 98.0+%, TCI America™

Catalog No. B36011G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B36011G Supplier TCI America Supplier No. B36011G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 99365-40-9
Molecular Formula C8H6BrNO
Molecular Weight (g/mol) 212.05
MDL Number MFCD02179605,MFCD22576660
InChI Key JARRYVQFBQVOBE-UHFFFAOYSA-N
Synonym 6-bromooxindole, 6-bromoindolin-2-one, 6-bromo-2-oxindole, 6-bromo-1,3-dihydro-2h-indol-2-one, 6-bromo-2-oxyindole, 6-bromo-1,3-dihydro-indol-2-one, 6-bromo-2-indolinone, 6-bromooxindol, 6-bromo-2,3-dihydro-1h-indol-2-one, 2h-indol-2-one, 6-bromo-1,3-dihydro
PubChem CID 2773289
IUPAC Name 6-bromo-2,3-dihydro-1H-indol-2-one
SMILES BrC1=CC=C2CC(=O)NC2=C1

Specifications

Melting Point 218°C
Quantity 1 g
Formula Weight 212.05
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromooxindole
TSCA No
Recommended Storage Refrigerator