missing translation for 'onlineSavingsMsg'
Learn More

6-Bromooxindole, 97%

Catalog No. AC399435000
Change view
Click to view available options
Quantity:
500 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC399435000 500 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC399435000 Supplier Thermo Scientific Chemicals Supplier No. 399435000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 99365-40-9 | C8H6BrNO | 212.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99365-40-9
Molecular Formula C8H6BrNO
Molecular Weight (g/mol) 212.05
MDL Number MFCD02179605,MFCD22576660
InChI Key JARRYVQFBQVOBE-UHFFFAOYSA-N
Synonym 6-bromooxindole, 6-bromoindolin-2-one, 6-bromo-2-oxindole, 6-bromo-1,3-dihydro-2h-indol-2-one, 6-bromo-2-oxyindole, 6-bromo-1,3-dihydro-indol-2-one, 6-bromo-2-indolinone, 6-bromooxindol, 6-bromo-2,3-dihydro-1h-indol-2-one, 2h-indol-2-one, 6-bromo-1,3-dihydro
PubChem CID 2773289
IUPAC Name 6-bromo-1,3-dihydroindol-2-one
SMILES BrC1=CC=C2CC(=O)NC2=C1

Specifications

Melting Point 217.0°C to 221.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 500 mg
Formula Weight 212.04
Percent Purity 97%
Chemical Name or Material 6-Bromooxindole
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only