missing translation for 'onlineSavingsMsg'
Learn More

6-Bromooxindole, 97%

Catalog No. p-7037302
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5601203 1 g
AAH5601206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5601203 Supplier Thermo Scientific Chemicals Supplier No. H5601203

CAS: 99365-40-9 | C8H6BrNO | 212.05 g/mol

6-Bromooxindole derivative is used in the preparation of p38alpha inhibitors as potential anti- inflammatories. It is used as an intermediate in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromooxindole derivative is used in the preparation of p38alpha inhibitors as potential anti- inflammatories. It is used as an intermediate in pharmaceuticals.

Solubility
Slightly soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 99365-40-9
Molecular Formula C8H6BrNO
Molecular Weight (g/mol) 212.05
MDL Number MFCD02179605,MFCD22576660
InChI Key JARRYVQFBQVOBE-UHFFFAOYSA-N
Synonym 6-bromooxindole, 6-bromoindolin-2-one, 6-bromo-2-oxindole, 6-bromo-1,3-dihydro-2h-indol-2-one, 6-bromo-2-oxyindole, 6-bromo-1,3-dihydro-indol-2-one, 6-bromo-2-indolinone, 6-bromooxindol, 6-bromo-2,3-dihydro-1h-indol-2-one, 2h-indol-2-one, 6-bromo-1,3-dihydro
PubChem CID 2773289
IUPAC Name 6-bromo-1,3-dihydroindol-2-one
SMILES BrC1=CC=C2CC(=O)NC2=C1

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 212.05
Percent Purity 97%
Chemical Name or Material 6-Bromooxindole
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only