missing translation for 'onlineSavingsMsg'
Learn More

6-Bromonicotinic Acid 98.0+%, TCI America™

Catalog No. p-7119863
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
B29615G 5 g
B296125G 25 g
2 options

Catalog No. B29615G

Supplier: TCI America B29615G

Only null left

Chemical Identifiers

CAS 6311-35-9
Molecular Formula C6H4BrNO2
Molecular Weight (g/mol) 202.01
MDL Number MFCD01927100
InChI Key JDJBRMNTXORYEN-UHFFFAOYSA-N
Synonym 6-bromonicotinic acid, 6-bromo nicotinic acid, 2-bromo-5-pyridinecarboxylic acid, 2-bromopyridine-5-carboxylic acid, 6-bromo-3-pyridine carboxylic acid, 6-bromonicotinicacid, 6-bromo-nicotinic acid, 6-bromo-3-pyridinecarboxylic acid, 2-bromo-5-pyridine carboxylic acid, 3-pyridinecarboxylic acid, 6-bromo
PubChem CID 238932
IUPAC Name 6-bromopyridine-3-carboxylic acid
SMILES OC(=O)C1=CC=C(Br)N=C1

Specifications

Melting Point 190°C
Color White-Yellow
Quantity 5 g
Formula Weight 202.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromonicotinic Acid