accessibility menu, dialog, popup

UserName

HOOBrN

CAS RN 6311-35-9

IUPAC Name: 6-bromopyridine-3-carboxylic acid
Synonyms: 6-bromopyridine-3-carboxylic acid
Molecular Weight (g/mol): 202.01
Molecular Formula: C6H4BrNO2
InChi Key: JDJBRMNTXORYEN-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(Br)N=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)