missing translation for 'onlineSavingsMsg'
Learn More

6-Bromonicotinic acid, 96%

Catalog No. p-4228463
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC435400010 1 g Glass bottle
AC435400100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC435400010 Supplier Thermo Scientific Chemicals Supplier No. 435400010

CAS: 6311-35-9 | C6H4BrNO2 | 202.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6311-35-9
Molecular Formula C6H4BrNO2
Molecular Weight (g/mol) 202.01
MDL Number MFCD01927100
InChI Key JDJBRMNTXORYEN-UHFFFAOYSA-N
Synonym 6-bromonicotinic acid, 6-bromo nicotinic acid, 2-bromo-5-pyridinecarboxylic acid, 2-bromopyridine-5-carboxylic acid, 6-bromo-3-pyridine carboxylic acid, 6-bromonicotinicacid, 6-bromo-nicotinic acid, 6-bromo-3-pyridinecarboxylic acid, 2-bromo-5-pyridine carboxylic acid, 3-pyridinecarboxylic acid, 6-bromo
PubChem CID 238932
IUPAC Name 6-bromopyridine-3-carboxylic acid
SMILES OC(=O)C1=CC=C(Br)N=C1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 179°C to 181°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 202.01
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromonicotinic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES.

RUO – Research Use Only