missing translation for 'onlineSavingsMsg'
Learn More

6-Bromoindole 98.0+%, TCI America™

Catalog No. B17945G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17945G 5 g
B17941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B17945G Supplier TCI America Supplier No. B17945G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 52415-29-9
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.047
MDL Number MFCD00238550
InChI Key MAWGHOPSCKCTPA-UHFFFAOYSA-N
Synonym 6-bromoindole, 1h-indole, 6-bromo, bromoindole-6, 6-bromo-indole, 6-brom-1h-indole, pubchem1662, 6-bromo-1-h-indole, 6-bromindol, maybridge1_006369
PubChem CID 676493
IUPAC Name 6-bromo-1H-indole
SMILES C1=CC(=CC2=C1C=CN2)Br

Specifications

Melting Point 95°C
Quantity 5 g
Formula Weight 196.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromoindole