missing translation for 'onlineSavingsMsg'
Learn More

6-Bromoindole, 98%, Thermo Scientific Chemicals

Catalog No. p-7047140
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL17703MD 250 mg
AAL1770303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL17703MD Supplier Thermo Scientific Chemicals Supplier No. L17703MD
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52415-29-9
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.047
MDL Number MFCD00238550
InChI Key MAWGHOPSCKCTPA-UHFFFAOYSA-N
Synonym 6-bromoindole, 1h-indole, 6-bromo, bromoindole-6, 6-bromo-indole, 6-brom-1h-indole, pubchem1662, 6-bromo-1-h-indole, 6-bromindol, maybridge1_006369
PubChem CID 676493
IUPAC Name 6-bromo-1H-indole
SMILES C1=CC(=CC2=C1C=CN2)Br

Specifications

Melting Point 91°C to 93°C
Boiling Point 70°C to 75°C (0.01 mmHg)
Quantity 250 mg
Beilstein 112711
Formula Weight 196.05
Percent Purity 98%
Chemical Name or Material 6-Bromoindole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only