Learn More
6-Bromoindole, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 397362500
| Quantity | 250mg |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 52415-29-9 | |
| 196.05 | |
| MAWGHOPSCKCTPA-UHFFFAOYSA-N | |
| 676493 | |
| C1=CC(=CC2=C1C=CN2)Br |
| C8H6BrN | |
| MFCD00238550 | |
| 6-bromoindole, 1h-indole, 6-bromo, bromoindole-6, 6-bromo-indole, 6-brom-1h-indole, pubchem1662, 6-bromo-1-h-indole, 6-bromindol, maybridge1_006369 | |
| 6-bromo-1H-indole |
Specifications
| 6-Bromoindole | |
| 95.0 | |
| Authentic | |
| Glass bottle | |
| 250mg | |
| 6-bromoindole, 1h-indole, 6-bromo, bromoindole-6, 6-bromo-indole, 6-brom-1h-indole, pubchem1662, 6-bromo-1-h-indole, 6-bromindol, maybridge1_006369 | |
| MAWGHOPSCKCTPA-UHFFFAOYSA-N | |
| 6-bromo-1H-indole | |
| 676493 | |
| 96% |
| 52415-29-9 | |
| 100.0 | |
| 95% min. (GC) | |
| C8H6BrN | |
| MFCD00238550 | |
| Solubility in water: insoluble | |
| C1=CC(=CC2=C1C=CN2)Br | |
| 196.05 | |
| 196.05 |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning