missing translation for 'onlineSavingsMsg'
Learn More

6-Bromoindole, 96%

Catalog No. AC397360050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC397360050 5 g Glass bottle
AC397360010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC397360050 Supplier Thermo Scientific Chemicals Supplier No. 397360050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 52415-29-9 | C8H6BrN | 196.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52415-29-9
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.05
MDL Number MFCD00238550
InChI Key MAWGHOPSCKCTPA-UHFFFAOYSA-N
Synonym 6-bromoindole, 1h-indole, 6-bromo, bromoindole-6, 6-bromo-indole, 6-brom-1h-indole, pubchem1662, 6-bromo-1-h-indole, 6-bromindol, maybridge1_006369
PubChem CID 676493
IUPAC Name 6-bromo-1H-indole
SMILES C1=CC(=CC2=C1C=CN2)Br

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 92.0°C to 96.0°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 196.05
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 6-Bromoindole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

RUO – Research Use Only