missing translation for 'onlineSavingsMsg'
Learn More

6-Bromoindole-2-carboxylic Acid 97.0+%, TCI America™

Catalog No. B43345G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43345G 5 g
B43341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43345G Supplier TCI America Supplier No. B43345G

Chemical Identifiers

CAS 16732-65-3
Molecular Formula C9H6BrNO2
Molecular Weight (g/mol) 240.056
MDL Number MFCD02664469
InChI Key SVBVYRYROZWKNJ-UHFFFAOYSA-N
Synonym 6-bromoindole-2-carboxylic acid, 6-bromo-2-indolecarboxylic acid, 1h-indole-2-carboxylic acid, 6-bromo, 6-bromoindole-2-carboxylicacid, zlchem 475, pubchem1664, acmc-1bqkh, intermediates-zcf02255, evoblocks eb21020, timtec-bb sbb014280
PubChem CID 4011696
IUPAC Name 6-bromo-1H-indole-2-carboxylic acid
SMILES C1=CC2=C(C=C1Br)NC(=C2)C(=O)O

Specifications

Melting Point 224°C
Quantity 5 g
Formula Weight 240.06
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromoindole-2-carboxylic Acid