accessibility menu, dialog, popup

UserName

BrHNOHO

CAS RN 16732-65-3

IUPAC Name: 6-bromo-1H-indole-2-carboxylic acid
Synonyms: 6-bromo-1H-indole-2-carboxylic acid
Molecular Weight (g/mol): 240.06
Molecular Formula: C9H6BrNO2
InChi Key: SVBVYRYROZWKNJ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=CC=C(Br)C=C2N1
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)