missing translation for 'onlineSavingsMsg'
Learn More

6-Bromoindirubin-3'-oxime, 97%

Catalog No. p-7057753
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67275MA 10 mg
AAJ67275MF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ67275MA Supplier Thermo Scientific Chemicals Supplier No. J67275MA
Only null left

CAS: 667463-62-9 | C16H10BrN3O2 | 356.179 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 667463-62-9
Molecular Formula C16H10BrN3O2
Molecular Weight (g/mol) 356.179
MDL Number MFCD08705318
InChI Key WNWSUJQVZJJGLF-UHFFFAOYSA-N
Synonym bio, 6-bromo-3-3-hydroxyamino indol-2-ylidene-1h-indol-2-one, z,3e-6'-bromo-3-hydroxyimino-1h,1'h-2,3'-biindolyliden-2'-one, 2h-indol-2-one, 6-bromo-3-3e-1,3-dihydro-3-hydroxyimino-2h-indol-2-ylidene-1,3-dihydro-, 3z, 6-bromo-3-3-hydroxyamino indol-2-ylidene indolin-2-one, 2z,3e-6'-bromo-3-hydroxyimino-2,3'-biindolinylidene-2'-one, 6-bromo-3-3-hydroxyamino-2h-indol-2-ylidene-1,3-dihydro-2h-indol-2-one
PubChem CID 24906282
IUPAC Name 6-bromo-3-[3-(hydroxyamino)indol-2-ylidene]-1H-indol-2-one
SMILES C1=CC2=C(C(=C3C4=C(C=C(C=C4)Br)NC3=O)N=C2C=C1)NO

Specifications

Color Red
Quantity 10 mg
Sensitivity Air and light sensitive
Formula Weight 356.17
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 6-Bromoindirubin-3'-oxime
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber NM3241900
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only