missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Bromocoumarin-3-carboxylic Acid 98.0+%, TCI America™
Supplier: TCI America B469625G
Specifications
| 6-Bromocoumarin-3-carboxylic Acid | |
| 193°C | |
| C10H5BrO4 | |
| 25 g | |
| XFQHPAXNKDYMOX-UHFFFAOYSA-N | |
| 6-bromo-2-oxochromene-3-carboxylic acid | |
| 694463 | |
| ≥98.0% (HPLC) |
| 2199-87-3 | |
| White-Yellow | |
| MFCD00047640 | |
| 6-bromo-2-oxo-2h-chromene-3-carboxylic acid, 6-bromocoumarin-3-carboxylic acid, acmc-20alzr, maybridge1_000837, 6-bromo-3-carboxy-coumarin, 6-bromo-coumarin-3-carboxylic acid, 2-oxo-6-bromo-2h-1-benzopyran-3-carboxylic acid, 2h-1-benzopyran-3-carboxylicacid,6-bromo-2-oxo, 2h-1-benzopyran-3-carboxylicacid, 6-bromo-2-oxo | |
| C1=CC2=C(C=C1Br)C=C(C(=O)O2)C(=O)O | |
| 269.05 | |
| 269.05 | |
| Crystalline Powder |
Chemical Identifiers
| 2199-87-3 | |
| 269.05 | |
| XFQHPAXNKDYMOX-UHFFFAOYSA-N | |
| 6-bromo-2-oxochromene-3-carboxylic acid |
| C10H5BrO4 | |
| MFCD00047640 | |
| 694463 | |
| C1=CC2=C(C=C1Br)C=C(C(=O)O2)C(=O)O |
Safety and Handling
TSCA : No