missing translation for 'onlineSavingsMsg'
Learn More

6-Bromocoumarin-3-carboxylic Acid 98.0+%, TCI America™

Catalog No. p-7123693
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
B46965G 5 g
B469625G 25 g
2 options

Catalog No. B46965G

Supplier: TCI America B46965G

Only null left

Chemical Identifiers

CAS 2199-87-3
Molecular Formula C10H5BrO4
Molecular Weight (g/mol) 269.05
MDL Number MFCD00047640
InChI Key XFQHPAXNKDYMOX-UHFFFAOYSA-N
PubChem CID 694463
IUPAC Name 6-bromo-2-oxochromene-3-carboxylic acid
SMILES C1=CC2=C(C=C1Br)C=C(C(=O)O2)C(=O)O

Specifications

Melting Point 193°C
Color White-Yellow
Quantity 5 g
Synonym 6-bromo-2-oxo-2h-chromene-3-carboxylic acid, 6-bromocoumarin-3-carboxylic acid, acmc-20alzr, maybridge1_000837, 6-bromo-3-carboxy-coumarin, 6-bromo-coumarin-3-carboxylic acid, 2-oxo-6-bromo-2h-1-benzopyran-3-carboxylic acid, 2h-1-benzopyran-3-carboxylicacid,6-bromo-2-oxo, 2h-1-benzopyran-3-carboxylicacid, 6-bromo-2-oxo
Formula Weight 269.05
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromocoumarin-3-carboxylic Acid