missing translation for 'onlineSavingsMsg'
Learn More

6-Bromochromone 98.0+%, TCI America™

Numéro de catalogue. B33355G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
B33355G 5 g
B333525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. B33355G Fournisseur TCI America Code fournisseur B33355G
Il en reste null

Identifiants chimiques

CAS 51483-92-2
Molecular Formula C9H5BrO2
Molecular Weight (g/mol) 225.041
MDL Number MFCD00239369
InChI Key XVNBWGGBXOJIDR-UHFFFAOYSA-N
Synonym 6-bromochromone, 6-bromo-4h-chromen-4-one, 4h-1-benzopyran-4-one,6-bromo, 4h-1-benzopyran-4-one, 6-bromo, acmc-1aser, 6-bromanylchromen-4-one, 6-bromo-1-benzopyran-4-one, 6-bromo-4h-chromen-4-one #, 6-bromo-4-oxo-4h-1-benzopyran
PubChem CID 548522
IUPAC Name 6-bromochromen-4-one
SMILES C1=CC2=C(C=C1Br)C(=O)C=CO2

Spécifications

Melting Point 138°C
Color White-Yellow
Quantity 5 g
Formula Weight 225.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromochromone