missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Bromo-o-cresol 94.0+%, TCI America™
$329.94 - $1159.44
Chemical Identifiers
| CAS | 13319-71-6 |
|---|---|
| Molecular Formula | C7H7BrO |
| Molecular Weight (g/mol) | 187.04 |
| MDL Number | MFCD09907876 |
| InChI Key | YXZPTVOCJLCMRO-UHFFFAOYSA-N |
| Synonym | 2-Bromo-6-methylphenol |
| PubChem CID | 11095329 |
| IUPAC Name | 2-bromo-6-methylphenol |
| SMILES | CC1=C(O)C(Br)=CC=C1 |
Chemical Identifiers
| 13319-71-6 | |
| 187.04 | |
| YXZPTVOCJLCMRO-UHFFFAOYSA-N | |
| 11095329 | |
| CC1=C(O)C(Br)=CC=C1 |
| C7H7BrO | |
| MFCD09907876 | |
| 2-Bromo-6-methylphenol | |
| 2-bromo-6-methylphenol |
Specifications
| 13319-71-6 | |
| Yellow | |
| C7H7BrO | |
| 5 g | |
| YXZPTVOCJLCMRO-UHFFFAOYSA-N | |
| 2-bromo-6-methylphenol | |
| 11095329 | |
| ≥94.0% (GC) | |
| 6-Bromo-o-cresol |
| 16°C | |
| 208°C | |
| MFCD09907876 | |
| 2-Bromo-6-methylphenol | |
| CC1=C(O)C(Br)=CC=C1 | |
| 187.04 | |
| 187.04 | |
| Liquid |
Safety and Handling
TSCA : No