missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-7-azaindole, 96%

Catalog No. AAH6686303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6686303 1 g
AAH6686306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6686303 Supplier Thermo Scientific Chemicals Supplier No. H6686303

CAS: 143468-13-7 | C7H5BrN2 | 197.035 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 143468-13-7
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.035
MDL Number MFCD06659667
InChI Key LKXJGVGBEDEAAW-UHFFFAOYSA-N
PubChem CID 11159915
IUPAC Name 6-bromo-1H-pyrrolo[2,3-b]pyridine
SMILES C1=CC(=NC2=C1C=CN2)Br

Specifications

Melting Point 192°C to 195°C
Color White to Yellow
Quantity 1 g
UN Number UN2811
Beilstein 5424619
Synonym 6-bromo-1h-pyrrolo 2,3-b pyridine, 6-bromo-7-azaindole, 1h-pyrrolo 2,3-b pyridine, 6-bromo, pubchem18671, acmc-209zuq, ksc493s6b, 1h-pyrrolo 2,3-b pyridine,6-bromo, 6-bromanyl-1h-pyrrolo 2,3-b pyridine
Formula Weight 197.04
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 6-Bromo-7-azaindole
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only