accessibility menu, dialog, popup

UserName

NHNBr

CAS RN 143468-13-7

IUPAC Name: 6-bromo-1H-pyrrolo[2,3-b]pyridine
Synonyms: 6-bromo-1H-pyrrolo[2,3-b]pyridine
Molecular Weight (g/mol): 197.04
Molecular Formula: C7H5BrN2
InChi Key: LKXJGVGBEDEAAW-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C=CNC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results