missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-4-methylquinoline, 96%

Catalog No. p-7031991
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH33298MD 250 mg
AAH3329803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH33298MD Supplier Thermo Scientific Chemicals Supplier No. H33298MD
Only null left

CAS: 41037-28-9 | C10H8BrN | 222.085 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 41037-28-9
Molecular Formula C10H8BrN
Molecular Weight (g/mol) 222.085
MDL Number MFCD09258893
InChI Key XBZQSJLYRMZRSZ-UHFFFAOYSA-N
Synonym quinoline, 6-bromo-4-methyl, 6-bromo-4-methyl-quinoline, 6-bromo lepidine, 4-methyl-6-bromoquinoline, quinoline,6-bromo-4-methyl, 6-bromanyl-4-methyl-quinoline
PubChem CID 12475362
IUPAC Name 6-bromo-4-methylquinoline
SMILES CC1=C2C=C(C=CC2=NC=C1)Br

Specifications

Melting Point 94°C to 95°C
Quantity 250 mg
Sensitivity Air and light sensitive
Formula Weight 222.08
Percent Purity 96%
Chemical Name or Material 6-Bromo-4-methylquinoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only