missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-3-methyl-1,3-benzoxazol-2(3H)-one, 97%, Thermo Scientific™

Chemical Identifiers

CAS 67927-44-0
Molecular Formula C8H6BrNO2
Molecular Weight (g/mol) 228.05
MDL Number MFCD00694789
InChI Key OASANCDKMGODJO-UHFFFAOYSA-N
Synonym 6-bromo-3-methyl-2,3-dihydro-1,3-benzoxazol-2-one, 6-bromo-3-methylbenzo d oxazol-2 3h-one, 6-bromo-3-methyl-1,3-benzoxazol-2 3h-one, bromomethyldihydrobenzoxazolone, 6-bromo-n-methylbenzoxazolone, 6-bromo-3-methyl-2 3h-benzoxazolone, 6-bromo-3-methyl-3h-benzooxazol-2-one, 6-bromo-3-methyl-1?3-benzoxazol-2 3h-one
PubChem CID 379590
IUPAC Name 6-bromo-3-methyl-1,3-benzoxazol-2-one
SMILES CN1C2=C(C=C(C=C2)Br)OC1=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC432710010
SDS View Documents
Thermo Scientific Chemicals
432710010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC432710050
SDS View Documents
Thermo Scientific Chemicals
432710050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

67927-44-0
228.05
OASANCDKMGODJO-UHFFFAOYSA-N
379590
CN1C2=C(C=C(C=C2)Br)OC1=O
C8H6BrNO2
MFCD00694789
6-bromo-3-methyl-2,3-dihydro-1,3-benzoxazol-2-one, 6-bromo-3-methylbenzo d oxazol-2 3h-one, 6-bromo-3-methyl-1,3-benzoxazol-2 3h-one, bromomethyldihydrobenzoxazolone, 6-bromo-n-methylbenzoxazolone, 6-bromo-3-methyl-2 3h-benzoxazolone, 6-bromo-3-methyl-3h-benzooxazol-2-one, 6-bromo-3-methyl-1?3-benzoxazol-2 3h-one
6-bromo-3-methyl-1,3-benzoxazol-2-one

Specifications

67927-44-0
100.0
96% min. (GC)
C8H6BrNO2
1g
OASANCDKMGODJO-UHFFFAOYSA-N
6-bromo-3-methyl-1,3-benzoxazol-2-one
379590
97%
96.0
Authentic
Glass bottle
MFCD00694789
6-bromo-3-methyl-2,3-dihydro-1,3-benzoxazol-2-one, 6-bromo-3-methylbenzo d oxazol-2 3h-one, 6-bromo-3-methyl-1,3-benzoxazol-2 3h-one, bromomethyldihydrobenzoxazolone, 6-bromo-n-methylbenzoxazolone, 6-bromo-3-methyl-2 3h-benzoxazolone, 6-bromo-3-methyl-3h-benzooxazol-2-one, 6-bromo-3-methyl-1?3-benzoxazol-2 3h-one
CN1C2=C(C=C(C=C2)Br)OC1=O
228.05
228.05
6-Bromo-3-methyl-1, 3-benzoxazol-2(3H)-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only