accessibility menu, dialog, popup

UserName

CAS RN 67927-44-0

IUPAC Name: 6-bromo-3-methyl-2,3-dihydro-1,3-benzoxazol-2-one
Synonyms: 6-bromo-3-methyl-1,3-benzoxazol-2-one
Molecular Weight (g/mol): 228.05
Molecular Formula: C8H6BrNO2
InChi Key: OASANCDKMGODJO-UHFFFAOYSA-N
SMILES: CN1C(=O)OC2=CC(Br)=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results