missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-3-methyl-1,3-benzoxazol-2(3H)-one, 97%

Catalog No. AC432710050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432710050 5 g Glass bottle
AC432710010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC432710050 Supplier Thermo Scientific Chemicals Supplier No. 432710050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 67927-44-0 | C8H6BrNO2 | 228.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 67927-44-0
Molecular Formula C8H6BrNO2
Molecular Weight (g/mol) 228.05
MDL Number MFCD00694789
InChI Key OASANCDKMGODJO-UHFFFAOYSA-N
Synonym 6-bromo-3-methyl-2,3-dihydro-1,3-benzoxazol-2-one, 6-bromo-3-methylbenzo d oxazol-2 3h-one, 6-bromo-3-methyl-1,3-benzoxazol-2 3h-one, bromomethyldihydrobenzoxazolone, 6-bromo-n-methylbenzoxazolone, 6-bromo-3-methyl-2 3h-benzoxazolone, 6-bromo-3-methyl-3h-benzooxazol-2-one, 6-bromo-3-methyl-1?3-benzoxazol-2 3h-one
PubChem CID 379590
IUPAC Name 6-bromo-3-methyl-1,3-benzoxazol-2-one
SMILES CN1C2=C(C=C(C=C2)Br)OC1=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 149°C to 150°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 228.05
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 6-Bromo-3-methyl-1, 3-benzoxazol-2(3H)-one
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

RUO – Research Use Only