missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-3,4-dihydro-2H-1,4-benzothiazine, 97%

Catalog No. p-7041134
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH61089MD 250 mg
AAH6108903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH61089MD Supplier Thermo Scientific Chemicals Supplier No. H61089MD

CAS: 187604-75-7 | C8H8BrNS | 230.123 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 187604-75-7
Molecular Formula C8H8BrNS
Molecular Weight (g/mol) 230.123
MDL Number MFCD23379863
InChI Key AUIBYERXYVUBIL-UHFFFAOYSA-N
Synonym 6-bromo-3,4-dihydro-2h-benzo b 1,4 thiazine
PubChem CID 67172430
IUPAC Name 6-bromo-3,4-dihydro-2H-1,4-benzothiazine
SMILES C1CSC2=C(N1)C=C(C=C2)Br

Specifications

Quantity 250 mg
Formula Weight 230.12
Percent Purity 97%
Chemical Name or Material 6-Bromo-3,4-dihydro-2H-1,4-benzothiazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only