missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-tetralone, 98%

Catalog No. AC330770010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC330770010 1 g Glass bottle
AC330770050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC330770010 Supplier Thermo Scientific Chemicals Supplier No. 330770010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4133-35-1 | C10H9BrO | 225.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4133-35-1
Molecular Formula C10H9BrO
Molecular Weight (g/mol) 225.08
MDL Number MFCD00239388
InChI Key BYHKDUFPSJWJDI-UHFFFAOYSA-N
Synonym 6-bromo-2-tetralone, 6-bromo-3,4-dihydronaphthalen-2 1h-one, 6-bromo-1,2,3,4-tetrahydronaphthalen-2-one, bromotetralone, 6-bromo-beta-tetralone, 6-bromo-3,4-dihydro-2 1h-naphthalenone, 2 1h-naphthalenone, 6-bromo-3,4-dihydro, 6-bromo-1,3,4-trihydronaphthalen-2-one, 6-bromo 2-tetralone
PubChem CID 2733553
IUPAC Name 6-bromo-3,4-dihydro-1H-naphthalen-2-one
SMILES C1CC2=C(CC1=O)C=CC(=C2)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 70°C to 76°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 225.08
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromo-2-tetralone, 98%
Show More Show Less
Recommended Storage Refrigerator +4°C

RUO – Research Use Only