missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Bromo-2-pyridinecarboxylic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 189°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 202.01 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 6-Bromo-2-pyridinecarboxylic Acid |
Safety and Handling
| TSCA | No |