missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-pyridinecarboxylic Acid 98.0+%, TCI America™

Catalog No. B30241G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30241G 1 g
B30245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30241G Supplier TCI America Supplier No. B30241G
Only null left

Chemical Identifiers

CAS 21190-87-4
Molecular Formula C6H4BrNO2
Molecular Weight (g/mol) 202.007
MDL Number MFCD00093197
InChI Key XURXQNUIGWHWHU-UHFFFAOYSA-N
Synonym 6-bromopicolinic acid, 6-bromo-2-pyridinecarboxylic acid, 6-bromo-2-pyridine carboxylic acid, 2-pyridinecarboxylic acid, 6-bromo, 2-bromo pyridine-6-carboxylic acid, 2-bromopyridine-6-carboxylic acid, 2-bromo-6-pyridinecarboxylic acid, 6-bromo-pyridine-2-carboxylic acid, 6-bromopicolinicacid, pubchem2422
PubChem CID 593919
IUPAC Name 6-bromopyridine-2-carboxylic acid
SMILES C1=CC(=NC(=C1)Br)C(=O)O

Specifications

Melting Point 189°C
Color White
Quantity 1 g
Formula Weight 202.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromo-2-pyridinecarboxylic Acid