missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-naphthol, 97%

Catalog No. p-7024213
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1853509 10 g
AAA1853518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1853509 Supplier Thermo Scientific Chemicals Supplier No. A1853509

missing translation for 'validMainframeMsg'

CAS: 15231-91-1 | C10H7BrO | 223.069 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15231-91-1
Molecular Formula C10H7BrO
Molecular Weight (g/mol) 223.069
MDL Number MFCD00004081
InChI Key YLDFTMJPQJXGSS-UHFFFAOYSA-N
Synonym 6-bromo-2-naphthol, 2-naphthalenol, 6-bromo, 6-bromo-naphthalen-2-ol, 6-bromo-2-naphthalenol, bromo-6 naphtol-2, 6-bromo2-hydroxynaphthalene, unii-87f10ahz3o, bromo-6 naphtol-2 french, 2-naphthol, 6-bromo, 6-bromo-beta-naphthol
PubChem CID 27144
ChEBI CHEBI:34466
IUPAC Name 6-bromonaphthalen-2-ol
SMILES C1=CC2=C(C=CC(=C2)Br)C=C1O

Specifications

Melting Point 126°C to 129°C
Quantity 10 g
Beilstein 1100270
Formula Weight 223.08
Percent Purity 97%
Chemical Name or Material 6-Bromo-2-naphthol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 239-279-0
RTECSNumber QL3365000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only