missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-naphthol, 97%

Catalog No. p-3757077
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107170250 25 g Glass bottle
AC107171000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC107170250 Supplier Thermo Scientific Chemicals Supplier No. 107170250
Only null left

CAS: 15231-91-1 | C10H7BrO | 223.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15231-91-1
Molecular Formula C10H7BrO
Molecular Weight (g/mol) 223.07
MDL Number MFCD00004081
InChI Key YLDFTMJPQJXGSS-UHFFFAOYSA-N
Synonym 6-bromo-2-naphthol, 2-naphthalenol, 6-bromo, 6-bromo-naphthalen-2-ol, 6-bromo-2-naphthalenol, bromo-6 naphtol-2, 6-bromo2-hydroxynaphthalene, unii-87f10ahz3o, bromo-6 naphtol-2 french, 2-naphthol, 6-bromo, 6-bromo-beta-naphthol
PubChem CID 27144
ChEBI CHEBI:34466
IUPAC Name 6-bromonaphthalen-2-ol
SMILES C1=CC2=C(C=CC(=C2)Br)C=C1O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 125°C to 130°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula BrC10H6OH
Quantity 25 g
Beilstein 06, 651
Formula Weight 223.07
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 6-Bromo-2-naphthol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 239-279-

RUO – Research Use Only