missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-hydroxy-3-methoxybenzaldehyde 99.0+%, TCI America™

Catalog No. B14871G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14871G 1 g
B14875G 5 g
B148725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B14871G Supplier TCI America Supplier No. B14871G
Only null left

Chemical Identifiers

CAS 20035-41-0
Molecular Formula C8H7BrO3
Molecular Weight (g/mol) 231.05
MDL Number MFCD00046152
InChI Key JUPJZUTYDWXZAQ-UHFFFAOYSA-N
PubChem CID 262237
IUPAC Name 6-bromo-2-hydroxy-3-methoxybenzaldehyde
SMILES COC1=CC=C(Br)C(C=O)=C1O

Specifications

Melting Point 103°C
Color Yellow
Quantity 1 g
Formula Weight 231.05
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromo-2-hydroxy-3-methoxybenzaldehyde