missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-chlorobenzothiazole, 97%

Catalog No. p-7031030
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3196203 1 g
AAH3196206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3196203 Supplier Thermo Scientific Chemicals Supplier No. H3196203
Only null left

CAS: 80945-86-4 | C7H3BrClNS | 248.522 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80945-86-4
Molecular Formula C7H3BrClNS
Molecular Weight (g/mol) 248.522
MDL Number MFCD04971822
InChI Key IJQSMNIZBBEBKI-UHFFFAOYSA-N
PubChem CID 2049871
IUPAC Name 6-bromo-2-chloro-1,3-benzothiazole
SMILES C1=CC2=C(C=C1Br)SC(=N2)Cl

Specifications

Melting Point 98°C to 100°C
Quantity 1 g
Synonym 6-bromo-2-chlorobenzothiazole, 6-bromo-2-chlorobenzo d thiazole, 6-bromo-2-chloro-benzothiazole, 2-chloro-6-bromobenzothiazole, benzothiazole, 6-bromo-2-chloro, 6-bromo-2-chlorbenzothiazole, benzothiazole,6-bromo-2-chloro, zlchem 563, pubchem11132, acmc-1bkg3
Formula Weight 248.53
Percent Purity 97%
Chemical Name or Material 6-Bromo-2-chlorobenzothiazole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only