accessibility menu, dialog, popup

UserName

BrSNCl

CAS RN 80945-86-4

IUPAC Name: 6-bromo-2-chloro-1,3-benzothiazole
Synonyms: 6-bromo-2-chloro-1,3-benzothiazole
Molecular Weight (g/mol): 248.52
Molecular Formula: C7H3BrClNS
InChi Key: IJQSMNIZBBEBKI-UHFFFAOYSA-N
SMILES: ClC1=NC2=CC=C(Br)C=C2S1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)