missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2,3-difluorotoluene, 95%

Catalog No. AAH3294103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3294103 1 g
AAH3294106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3294103 Supplier Thermo Scientific Chemicals Supplier No. H3294103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 847502-81-2 | C7H5BrF2 | 207.018 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 847502-81-2
Molecular Formula C7H5BrF2
Molecular Weight (g/mol) 207.018
MDL Number MFCD07777161
InChI Key YXTCBTSPHSNDHD-UHFFFAOYSA-N
PubChem CID 12137677
IUPAC Name 1-bromo-3,4-difluoro-2-methylbenzene
SMILES CC1=C(C=CC(=C1F)F)Br

Specifications

Refractive Index 1.5105
Quantity 1 g
Synonym 6-bromo-2,3-difluorotoluene, 3,4-difluoro-2-methylbromobenzene, benzene,1-bromo-3,4-difluoro-2-methyl, benzene, 1-bromo-3,4-difluoro-2-methyl, 2-bromo-5,6-difluorotoluene, 3,4-difluoro-2-methyl bromobenzene, 3,4-difluoro-2-methyl-bromobenzene
Formula Weight 207.02
Percent Purity 97%
Chemical Name or Material 6-Bromo-2,3-difluorotoluene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only