missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2,3-difluorobenzaldehyde, 98%

Catalog No. p-7030953
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3185503 1 g
AAH3185506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3185503 Supplier Thermo Scientific Chemicals Supplier No. H3185503

CAS: 360576-04-1 | C7H3BrF2O | 221.001 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 360576-04-1
Molecular Formula C7H3BrF2O
Molecular Weight (g/mol) 221.001
MDL Number MFCD07368119
InChI Key LAVPYRPTHABUAD-UHFFFAOYSA-N
Synonym benzaldehyde, 6-bromo-2,3-difluoro, 6-bromo-2,3-difluoro-benzaldehyde, 2-bromo-5,6-difluorobenzaldehyde, benzaldehyde,6-bromo-2,3-difluoro
PubChem CID 9877851
IUPAC Name 6-bromo-2,3-difluorobenzaldehyde
SMILES C1=CC(=C(C(=C1F)F)C=O)Br

Specifications

Melting Point 39°C to 43°C
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 221
Percent Purity 98%
Chemical Name or Material 6-Bromo-2,3-difluorobenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only