missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2,2'-bipyridyl 97.0+%, TCI America™

Catalog No. B41141G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41141G 1 g
B41145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41141G Supplier TCI America Supplier No. B41141G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10495-73-5
Molecular Formula C10H7BrN2
Molecular Weight (g/mol) 235.08
MDL Number MFCD01318953
InChI Key NCRIDSGPLISUEU-UHFFFAOYSA-N
PubChem CID 11791152
IUPAC Name 6-bromo-2,2'-bipyridine
SMILES BrC1=CC=CC(=N1)C1=CC=CC=N1

Specifications

Melting Point 74°C
Color White-Yellow
Boiling Point 133°C
Quantity 1 g
Synonym 6-Bromo-2,2′C-bipyridine
Formula Weight 235.08
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromo-2,2′-bipyridyl