accessibility menu, dialog, popup

UserName

CAS RN 10495-73-5

IUPAC Name: 6-bromo-2,2'-bipyridine
Synonyms: 6-bromo-2,2'-bipyridine
Molecular Weight (g/mol): 235.08
Molecular Formula: C10H7BrN2
InChi Key: NCRIDSGPLISUEU-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=N1)C1=CC=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results