missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-1-tetralone 95.0+%, TCI America™

Catalog No. B48415G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48415G 5 g
B48411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48415G Supplier TCI America Supplier No. B48415G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 66361-67-9
Molecular Formula C10H9BrO
Molecular Weight (g/mol) 225.09
MDL Number MFCD04114378
InChI Key OSDHOOBPMBLALZ-UHFFFAOYSA-N
Synonym 6-bromo-1-tetralone, 6-bromo-3,4-dihydronaphthalen-1 2h-one, 6-bromotetral-1-one, 6-bromo-tetral-1-on, 6-bromotetralone, 1 2h-naphthalenone, 6-bromo-3,4-dihydro, 6-bromo-1,2,3,4-tetrahydronaphthalen-1-one, 6-bromo-3,4-dihydro-1 2h-naphthalenone, 6-bromoteralone, 6-bromo-tetralone
PubChem CID 10105069
IUPAC Name 6-bromo-1,2,3,4-tetrahydronaphthalen-1-one
SMILES BrC1=CC2=C(C=C1)C(=O)CCC2

Specifications

Melting Point 44°C
Color White-Yellow
Boiling Point 116°C
Quantity 5 g
Formula Weight 225.09
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 6-Bromo-1-tetralone