missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-1-hexene 95.0+%, TCI America™

Catalog No. p-7119483
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21065G 5 g
B210625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21065G Supplier TCI America Supplier No. B21065G
Only null left

Chemical Identifiers

CAS 2695-47-8
Molecular Formula C6H11Br
Molecular Weight (g/mol) 163.058
MDL Number MFCD00000269
InChI Key RIMXEJYJXDBLIE-UHFFFAOYSA-N
Synonym 6-bromo-1-hexene, 5-hexenyl bromide, 1-hexene, 6-bromo, 1-bromo-5-hexene, 6-bromohexene, 6-bromo-hexene, 6-bromo-hex-1-ene, l-bromo-5-hexene, 6-bromo-1-hexen, 1-bromohex-5-ene
PubChem CID 75906
IUPAC Name 6-bromohex-1-ene
SMILES C=CCCCCBr

Specifications

Color Yellow
Boiling Point 147°C
Quantity 5 g
UN Number 1993
Formula Weight 163.06
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 6-Bromo-1-hexene
EINECSNumber (2)-0067
TSCA Yes
Recommended Storage Freezer