missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-1-hexene, 97%

Catalog No. p-2690886
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC364700010 1 g Glass bottle
AC364700050 5 g Glass bottle
AC364700250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC364700010 Supplier Thermo Scientific Chemicals Supplier No. 364700010
Only null left

CAS: 2695-47-8 | C6H11Br | 163.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2695-47-8
Molecular Formula C6H11Br
Molecular Weight (g/mol) 163.06
MDL Number MFCD00000269
InChI Key RIMXEJYJXDBLIE-UHFFFAOYSA-N
Synonym 6-bromo-1-hexene, 5-hexenyl bromide, 1-hexene, 6-bromo, 1-bromo-5-hexene, 6-bromohexene, 6-bromo-hexene, 6-bromo-hex-1-ene, l-bromo-5-hexene, 6-bromo-1-hexen, 1-bromohex-5-ene
PubChem CID 75906
IUPAC Name 6-bromohex-1-ene
SMILES C=CCCCCBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 1.2170g/mL
Boiling Point 47.0°C to 51.0°C (16.0 mmHg)
Flash Point 52°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4645 to 1.4665 (20°C, 589nm)
Linear Formula Br(CH2)4CH=CH2
Quantity 1 g
Specific Gravity 1.217
Formula Weight 163.06
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 6-Bromo-1-hexene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
EINECSNumber 220-267-9

RUO – Research Use Only