Learn More
6-Bromo-1-hexene, 95%
CAS: 2695-47-8 | C6H11Br | 163.058 g/mol
Supplier: Thermo Scientific Chemicals L1290906
Description
6-Bromo-1-hexene was used in the preparation of epoxide, 6-bromo-1,2-epoxyhexane. It was also used in a simple, direct preparation of 4-alkenylbenzonitriles via anionic reduced forms of terephthalonitrile in liquid ammonia.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-Bromo-1-hexene was used in the preparation of epoxide, 6-bromo-1,2-epoxyhexane. It was also used in a simple, direct preparation of 4-alkenylbenzonitriles via anionic reduced forms of terephthalonitrile in liquid ammonia.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| 6-Bromo-1-hexene | |
| 1.217 | |
| 54°C (129°F) | |
| C6H11Br | |
| Br(CH2)4CH=CH2 | |
| 5 g | |
| 506203 | |
| Not miscible in water. | |
| C=CCCCCBr | |
| 163.058 | |
| 163.06 |
| 2695-47-8 | |
| 147°C | |
| Characteristic | |
| 1.4652 | |
| MFCD00000269 | |
| UN1993 | |
| 6-bromo-1-hexene, 5-hexenyl bromide, 1-hexene, 6-bromo, 1-bromo-5-hexene, 6-bromohexene, 6-bromo-hexene, 6-bromo-hex-1-ene, l-bromo-5-hexene, 6-bromo-1-hexen, 1-bromohex-5-ene | |
| RIMXEJYJXDBLIE-UHFFFAOYSA-N | |
| 6-bromohex-1-ene | |
| 75906 | |
| 95% |
Chemical Identifiers
| 2695-47-8 | |
| 163.058 | |
| RIMXEJYJXDBLIE-UHFFFAOYSA-N | |
| 75906 | |
| C=CCCCCBr |
| C6H11Br | |
| MFCD00000269 | |
| 6-bromo-1-hexene, 5-hexenyl bromide, 1-hexene, 6-bromo, 1-bromo-5-hexene, 6-bromohexene, 6-bromo-hexene, 6-bromo-hex-1-ene, l-bromo-5-hexene, 6-bromo-1-hexen, 1-bromohex-5-ene | |
| 6-bromohex-1-ene |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 220-267-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only