missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-1-hexene, 95%

Catalog No. AAL1290903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1290903 1 g
AAL1290906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1290903 Supplier Thermo Scientific Chemicals Supplier No. L1290903

CAS: 2695-47-8 | C6H11Br | 163.058 g/mol

6-Bromo-1-hexene was used in the preparation of epoxide, 6-bromo-1,2-epoxyhexane. It was also used in a simple, direct preparation of 4-alkenylbenzonitriles via anionic reduced forms of terephthalonitrile in liquid ammonia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromo-1-hexene was used in the preparation of epoxide, 6-bromo-1,2-epoxyhexane. It was also used in a simple, direct preparation of 4-alkenylbenzonitriles via anionic reduced forms of terephthalonitrile in liquid ammonia.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2695-47-8
Molecular Formula C6H11Br
Molecular Weight (g/mol) 163.058
MDL Number MFCD00000269
InChI Key RIMXEJYJXDBLIE-UHFFFAOYSA-N
Synonym 6-bromo-1-hexene, 5-hexenyl bromide, 1-hexene, 6-bromo, 1-bromo-5-hexene, 6-bromohexene, 6-bromo-hexene, 6-bromo-hex-1-ene, l-bromo-5-hexene, 6-bromo-1-hexen, 1-bromohex-5-ene
PubChem CID 75906
IUPAC Name 6-bromohex-1-ene
SMILES C=CCCCCBr

Specifications

Density 1.217
Boiling Point 147°C
Flash Point 54°C (129°F)
Odor Characteristic
Refractive Index 1.4652
Linear Formula Br(CH2)4CH=CH2
Quantity 1 g
UN Number UN1993
Beilstein 506203
Solubility Information Not miscible in water.
Formula Weight 163.06
Percent Purity 95%
Chemical Name or Material 6-Bromo-1-hexene
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 220-267-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only